missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl alpha-L-rhamnopyranoside, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2412906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesKeep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Methyl alpha-L-rhamnopyranoside | |
106°C to 108°C | |
MFCD00067654 | |
methyl alpha-l-rhamnopyranoside, methyl-alpha-l-rhamnopyranoside, 1-o-methyl-alpha-rhamnose, methyl 6-deoxy-alpha-l-mannopyranoside, methyl a-l-rhamnopyranoside, rao, methyl rhamnopyranoside, 2r,3r,4r,5r,6s-2-methoxy-6-methyloxane-3,4,5-triol, 2r,3r,4r,5r,6s-2-methoxy-6-methyltetrahydro-2h-pyran-3,4,5-triol, methyl-a-l-rhamnopyranoside | |
CC1C(C(C(C(O1)OC)O)O)O | |
178.184 | |
178.19 |
14917-55-6 | |
C7H14O5 | |
5 g | |
OHWCAVRRXKJCRB-PAMBMQIZSA-N | |
(2R,3R,4R,5R,6S)-2-methoxy-6-methyloxane-3,4,5-triol | |
84695 | |
98% |
Chemical Identifiers
14917-55-6 | |
178.184 | |
OHWCAVRRXKJCRB-PAMBMQIZSA-N | |
84695 | |
CC1C(C(C(C(O1)OC)O)O)O |
C7H14O5 | |
MFCD00067654 | |
methyl alpha-l-rhamnopyranoside, methyl-alpha-l-rhamnopyranoside, 1-o-methyl-alpha-rhamnose, methyl 6-deoxy-alpha-l-mannopyranoside, methyl a-l-rhamnopyranoside, rao, methyl rhamnopyranoside, 2r,3r,4r,5r,6s-2-methoxy-6-methyloxane-3,4,5-triol, 2r,3r,4r,5r,6s-2-methoxy-6-methyltetrahydro-2h-pyran-3,4,5-triol, methyl-a-l-rhamnopyranoside | |
(2R,3R,4R,5R,6S)-2-methoxy-6-methyloxane-3,4,5-triol |
Safety and Handling
EINECSNumber : 238-987-7
TSCA : No
RUO – Research Use Only