Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl alpha-D-galactopyranoside monohydrate

$20.20 - $57.57

Chemical Identifiers

Chemical Name or Material Methyl alpha-D-galactopyranoside monohydrate
CAS 34004-14-3
Molecular Formula C7H14O6·H2O
Formula Weight 212.20 (194.18 Anhydrous)
MDL Number MFCD00150748
Beilstein 81570
Synonym Methyl alpha-D-galactoside monohydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0804903

  SDS
alfa aesar™
L0804903
1g Each for $20.20

AAL0804906

  SDS
alfa aesar™
L0804906
5g Each for $57.57
Description & Specifications

Specifications

Formula Weight 212.20 (194.18 Anhydrous)
Chemical Name or Material Methyl alpha-D-galactopyranoside monohydrate
CAS 34004-14-3
Molecular Formula C7H14O6·H2O
MDL Number MFCD00150748
Beilstein 81570
Synonym Methyl alpha-D-galactoside monohydrate
Melting Point 105° to 108°C
Quantity 1g
Optical Rotation +169° (c=1.5 in Methanol)
Recommended Storage 2° to 8°C

Applications
Methyl alpha-D-galactopyranoside monohydrate is used in medicine, industry.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

2° to 8°C