Please login to your online account to display your discounted pricing

Methyl Alcohol , Macron Fine Chemicals™


macron

Quantity 4L
Packaging Glass

Manufacturer: avantor performance  300408

 View more versions of this product

Catalog No. MK30044

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 32.04
Quantity 4L
Name Note Anhydrous
Molecular Formula CH3OH
Packaging Glass
CAS 67-56-1
Chemical Name or Material Methyl Alcohol

Specifications

Chemical Name or Material Methyl Alcohol
Name Note Anhydrous
CAS 67-56-1
Molecular Formula CH3OH
Formula Weight 32.04