missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Alcohol , Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH3OH
Formula Weight 32.04
Chemical Name or Material Methyl Alcohol, Anhydrous
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK300420 Avantor Performance
300419
Lined Steel Dr 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK30044 Avantor Performance
300408
Glass 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

67-56-1
CH3OH
32.04
Methyl Alcohol, Anhydrous

Specifications

32.04
CH3OH
Lined Steel Dr
20L
67-56-1
Methyl Alcohol, Anhydrous