Please login to your online account to display your discounted pricing

Methyl Alcohol ACS AR, Macron Fine Chemicals™


macron

Quantity 4L
Packaging Glass

Manufacturer: avantor performance  301708

 View more versions of this product

Catalog No. MK30174

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Glass
Quantity 4L
Chemical Name or Material Methyl Alcohol
Name Note Anhydrous
CAS 67-56-1
Purity Grade ACS Reagent
Molecular Formula CH3OH
Formula Weight 32.04g/mol

Specifications

Chemical Name or Material Methyl Alcohol
Name Note Anhydrous
CAS 67-56-1
Purity Grade ACS Reagent
Molecular Formula CH3OH
Formula Weight 32.04g/mol