Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl acetopyruvate, 98%

$75.74 - $278.61

Chemical Identifiers

Chemical Name or Material Methyl acetopyruvate
CAS 20577-61-1
Molecular Formula C8H8O4
Formula Weight 144.13
MDL Number MFCD00043638
Synonym Acetopyruvic acid methyl ester; Methyl 2,4-dioxovalerate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1119006

 
alfa aesar™
A1119006
5g Each for $75.74

AAA1119014

 
alfa aesar™
A1119014
25g Each for $278.61
Description & Specifications

Specifications

Formula Weight 144.13
Chemical Name or Material Methyl acetopyruvate
Assay 98%
Molecular Formula C8H8O4
MDL Number MFCD00043638
CAS 20577-61-1
Melting Point 60° to 63°C
Synonym Acetopyruvic acid methyl ester; Methyl 2,4-dioxovalerate
Quantity 5g