missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 371830010

 View more versions of this product

Catalog No. AC371830010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl acetate
-98.0°C
0.9300g/mL
−10°C
99% min. (GC)
C3H6O2
CH3CO2CH3
1 L
0.0005 % max.
15, 6082
Solubility in water: 250g/L (20°C). Other solubilities: miscible with alcohol and ether
CC(=O)OC
74.08
0.38 mPa.s (20°C)
74.08
Extra Dry
79-20-9
Colorless to Yellow
57.4°C
Authentic
AcroSeal™ Glass bottle
1.3600 to 1.3620
MFCD00008711
02, 224
0.93
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
KXKVLQRXCPHEJC-UHFFFAOYSA-N
methyl acetate
6584
CHEBI:77700
99+%
Solution

Chemical Identifiers

79-20-9
74.08
KXKVLQRXCPHEJC-UHFFFAOYSA-N
6584
methyl acetate
C3H6O2
MFCD00008711
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
CHEBI:77700
CC(=O)OC
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-185-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only