Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 7-chloroheptanoate, 98%

$87.28 - $257.05

Chemical Identifiers

Chemical Name or Material Methyl 7-chloroheptanoate
CAS 26040-62-0
Molecular Formula C8ClH15O2
Linear Formula Cl(CH2)6CO2CH3
Formula Weight 178.66
MDL Number MFCD09751145
Synonym 7-Chloroheptanoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2709806

 
alfa aesar™
H2709806
5g Each for $87.28

AAH2709814

 
alfa aesar™
H2709814
25g Each for $257.05
Description & Specifications

Specifications

Formula Weight 178.66
Chemical Name or Material Methyl 7-chloroheptanoate
Assay 98%
Molecular Formula C8ClH15O2
Linear Formula Cl(CH2)6CO2CH3
MDL Number MFCD09751145
CAS 26040-62-0
Synonym 7-Chloroheptanoic acid methyl ester
Quantity 5g