missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-O-(N-heptylcarbamoyl)-alpha-D-glucopyranoside, 98%

$81.27 - $121.51

Chemical Identifiers

CAS 115457-83-5
Molecular Formula C15H29NO7
Molecular Weight (g/mol) 335.397
MDL Number MFCD00077425
InChI Key XPIVOYOQXKNYHA-RGDJUOJXSA-N
Synonym hecameg, a-d-glucopyranoside, 6-o-n-heptylcarbamoyl methylglucoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxytetrahydro-2h-pyran-2-yl methyl heptylcarbamate, methyl 6-o-n-heptylcarbamoyl-a-d-glucopyranoside, methyl 6-o-n-heptylcarbamoyl-alpha-d-glucopyranoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxyoxan-2-yl methyl n-heptylcarbamate, 6-o-n-heptylcarbamoyl methyl-alpha-d-glucopyranoside, 6-o-n-heptylcarbamoyl-methyl-alpha-d-glucopyranoside, a-d-glucopyranoside,methyl,6-heptylcarbamate, 6-o-n-heptylcarbamoyl-methyl-
PubChem CID 2733955
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl N-heptylcarbamate
SMILES CCCCCCCNC(=O)OCC1C(C(C(C(O1)OC)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66834ME View Documents Alfa Aesar™
J66834ME
500mg Each for $81.27
 
AAJ6683403 View Documents Alfa Aesar™
J6683403
1g Each for $121.51
 
Specifications

Chemical Identifiers

115457-83-5
335.397
XPIVOYOQXKNYHA-RGDJUOJXSA-N
2733955
CCCCCCCNC(=O)OCC1C(C(C(C(O1)OC)O)O)O
C15H29NO7
MFCD00077425
hecameg, a-d-glucopyranoside, 6-o-n-heptylcarbamoyl methylglucoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxytetrahydro-2h-pyran-2-yl methyl heptylcarbamate, methyl 6-o-n-heptylcarbamoyl-a-d-glucopyranoside, methyl 6-o-n-heptylcarbamoyl-alpha-d-glucopyranoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxyoxan-2-yl methyl n-heptylcarbamate, 6-o-n-heptylcarbamoyl methyl-alpha-d-glucopyranoside, 6-o-n-heptylcarbamoyl-methyl-alpha-d-glucopyranoside, a-d-glucopyranoside,methyl,6-heptylcarbamate, 6-o-n-heptylcarbamoyl-methyl-
[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl N-heptylcarbamate

Specifications

+101° (c=1 in Water)
100°C
115457-83-5
MFCD00077425
14,4621
XPIVOYOQXKNYHA-RGDJUOJXSA-N
[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl N-heptylcarbamate
2733955
Powder
Methyl 6-O-(N-heptylcarbamoyl)-alpha-D-glucopyranoside
White
500mg
C15H29NO7
3560078
hecameg, a-d-glucopyranoside, 6-o-n-heptylcarbamoyl methylglucoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxytetrahydro-2h-pyran-2-yl methyl heptylcarbamate, methyl 6-o-n-heptylcarbamoyl-a-d-glucopyranoside, methyl 6-o-n-heptylcarbamoyl-alpha-d-glucopyranoside, 2r,3s,4s,5r,6s-3,4,5-trihydroxy-6-methoxyoxan-2-yl methyl n-heptylcarbamate, 6-o-n-heptylcarbamoyl methyl-alpha-d-glucopyranoside, 6-o-n-heptylcarbamoyl-methyl-alpha-d-glucopyranoside, a-d-glucopyranoside,methyl,6-heptylcarbamate, 6-o-n-heptylcarbamoyl-methyl-
CCCCCCCNC(=O)OCC1C(C(C(C(O1)OC)O)O)O
335.397
335.39
98%
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C