missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-methylnicotinate, 97+%

Manufacturer:  Alfa Aesar™ L1227603

 View more versions of this product

Catalog No. AAL1227603



Specifications

Specifications

Methyl 6-methylnicotinate
152°C to 154°C (40mmHg)
1g
C8H9NO2
122940
VYPPZXZHYDSBSJ-UHFFFAOYSA-N
methyl 6-methylpyridine-3-carboxylate
231548
≥97%
103°C (217°F)
31°C to 36°C
5470-70-2
MFCD00006340
methyl 6-methylnicotinate, methyl-6-methylnicotinate, 6-methylnicotinic acid methyl ester, etoricoxib impurity e, unii-ya0k0cm5pk, methyl 6-methyl nicotinate, 6-methyl-nicotinic acid methyl ester, 6-methyl nicotinic acid methyl ester, ya0k0cm5pk, 3-methoxycarbonyl-6-methylpyridine
CC1=NC=C(C=C1)C(=O)OC
151.165
151.17

Chemical Identifiers

5470-70-2
151.165
VYPPZXZHYDSBSJ-UHFFFAOYSA-N
231548
CC1=NC=C(C=C1)C(=O)OC
C8H9NO2
MFCD00006340
methyl 6-methylnicotinate, methyl-6-methylnicotinate, 6-methylnicotinic acid methyl ester, etoricoxib impurity e, unii-ya0k0cm5pk, methyl 6-methyl nicotinate, 6-methyl-nicotinic acid methyl ester, 6-methyl nicotinic acid methyl ester, ya0k0cm5pk, 3-methoxycarbonyl-6-methylpyridine
methyl 6-methylpyridine-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents