Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-methylnicotinate, 97+%

$158.22 - $552.99

Chemical Identifiers

Chemical Name or Material Methyl 6-methylnicotinate
CAS 5470-70-2
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00006340
Beilstein 122940
Synonym Methyl 6-methylpyridine-3-carboxylate; 6-Methylnicotinic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1227606

  SDS
alfa aesar™
L1227606
5g Each for $158.22

AAL1227614

  SDS
alfa aesar™
L1227614
25g Each for $552.99
Description & Specifications

Specifications

Formula Weight 151.17
CAS 5470-70-2
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Quantity 5g
MDL Number MFCD00006340
Molecular Formula C8H9NO2
Melting Point 31° to 36°C
Synonym Methyl 6-methylpyridine-3-carboxylate; 6-Methylnicotinic acid methyl ester
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Beilstein 122940
Flash Point 103°C (217°F)
Boiling Point 152° to 154°C (40mmHg)
Assay ≥97%
Chemical Name or Material Methyl 6-methylnicotinate
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.