Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-methylnicotinate, 97+%


Quantity 25g

Manufacturer: alfa aesar™  L1227614

 View more versions of this product

Catalog No. AAL1227614

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 151.17
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Chemical Name or Material Methyl 6-methylnicotinate
Molecular Formula C8H9NO2
Beilstein 122940
Assay ≥97%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
CAS 5470-70-2
Flash Point 103°C (217°F)
Boiling Point 152° to 154°C (40mmHg)
MDL Number MFCD00006340
Synonym Methyl 6-methylpyridine-3-carboxylate; 6-Methylnicotinic acid methyl ester
Quantity 25g
Melting Point 31° to 36°C

Specifications

Chemical Name or Material Methyl 6-methylnicotinate
CAS 5470-70-2
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00006340
Beilstein 122940
Synonym Methyl 6-methylpyridine-3-carboxylate; 6-Methylnicotinic acid methyl ester
Accessories

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents