Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-methoxyindole-2-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 6-methoxyindole-2-carboxylate
CAS 98081-83-5
Formula Weight 205.21
MDL Number MFCD00134301
Beilstein 395862
Synonym 6-Methoxy-indole-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6652803

 
alfa aesar™
H6652803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6652806

 
alfa aesar™
H6652806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.21
Melting Point 117° to 119°C
Physical Form Solid
Assay 97%
MDL Number MFCD00134301
CAS 98081-83-5
Beilstein 395862
Synonym 6-Methoxy-indole-2-carboxylic acid methyl ester
Quantity 1g
Chemical Name or Material Methyl 6-methoxyindole-2-carboxylate