missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-(hydroxymethyl)pyridine-2-carboxylate, 95%

Manufacturer:  Alfa Aesar™ H6379306

 View more versions of this product

Catalog No. AAH6379306



Specifications

Specifications

Methyl 6-(hydroxymethyl)pyridine-2-carboxylate
5g
C8H9NO3
methyl 6-hydroxymethyl picolinate, methyl 6-hydroxymethyl pyridine-2-carboxylate, methyl-6-hydroxymethyl-2-carboxylate pyridine, 6-hydroxymethyl-2-pyridinecarboxylic acid methyl ester, 6-hydroxymethyl-pyridine-2-carboxylic acid methyl ester, methyl 6-hydroxymethyl-2-pyridinecarboxylate, methyl6-hydroxymethyl picolinate, 2-pyridinecarboxylic acid, 6-hydroxymethyl-, methyl ester, methyl 6-hydroxymethyl-2-pyridine carboxylic acid, 2-pyridinecarboxylicacid, 6-hydroxymethyl-, methyl ester
COC(=O)C1=NC(=CC=C1)CO
167.164
167.16
95%
White
39977-44-1
MFCD03840850
DVIUNMLAPDJWHL-UHFFFAOYSA-N
methyl 6-(hydroxymethyl)pyridine-2-carboxylate
11513809
Solid

Chemical Identifiers

39977-44-1
167.164
DVIUNMLAPDJWHL-UHFFFAOYSA-N
11513809
COC(=O)C1=NC(=CC=C1)CO
C8H9NO3
MFCD03840850
methyl 6-hydroxymethyl picolinate, methyl 6-hydroxymethyl pyridine-2-carboxylate, methyl-6-hydroxymethyl-2-carboxylate pyridine, 6-hydroxymethyl-2-pyridinecarboxylic acid methyl ester, 6-hydroxymethyl-pyridine-2-carboxylic acid methyl ester, methyl 6-hydroxymethyl-2-pyridinecarboxylate, methyl6-hydroxymethyl picolinate, 2-pyridinecarboxylic acid, 6-hydroxymethyl-, methyl ester, methyl 6-hydroxymethyl-2-pyridine carboxylic acid, 2-pyridinecarboxylicacid, 6-hydroxymethyl-, methyl ester
methyl 6-(hydroxymethyl)pyridine-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents