missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 6-Fluoronicotinate 98.0+%, TCI America™
$150.97 - $499.37
Chemical Identifiers
CAS | 1427-06-1 |
---|---|
Molecular Formula | C7H6FNO2 |
Molecular Weight (g/mol) | 155.128 |
MDL Number | MFCD03095098 |
InChI Key | HLYBWNNPVXFCPZ-UHFFFAOYSA-N |
Synonym | methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate |
PubChem CID | 13758765 |
IUPAC Name | methyl 6-fluoropyridine-3-carboxylate |
SMILES | COC(=O)C1=CN=C(C=C1)F |
Chemical Identifiers
1427-06-1 | |
155.128 | |
HLYBWNNPVXFCPZ-UHFFFAOYSA-N | |
13758765 | |
COC(=O)C1=CN=C(C=C1)F |
C7H6FNO2 | |
MFCD03095098 | |
methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate | |
methyl 6-fluoropyridine-3-carboxylate |
Specifications
1427-06-1 | |
White-Yellow | |
MFCD03095098 | |
methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate | |
COC(=O)C1=CN=C(C=C1)F | |
155.128 | |
155.13 | |
Crystalline Powder |
51°C | |
C7H6FNO2 | |
1 g | |
HLYBWNNPVXFCPZ-UHFFFAOYSA-N | |
methyl 6-fluoropyridine-3-carboxylate | |
13758765 | |
≥98.0% (GC) | |
Methyl 6-Fluoronicotinate |
Safety and Handling
TSCA : No