missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 6-fluoronicotinate, 95%, Thermo Scientific Chemicals
$115.56 - $369.90
Chemical Identifiers
CAS | 1427-06-1 |
---|---|
Molecular Formula | C7H6FNO2 |
Molecular Weight (g/mol) | 155.128 |
MDL Number | MFCD03095098 |
InChI Key | HLYBWNNPVXFCPZ-UHFFFAOYSA-N |
PubChem CID | 13758765 |
IUPAC Name | methyl 6-fluoropyridine-3-carboxylate |
SMILES | COC(=O)C1=CN=C(C=C1)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH61302MD
|
Thermo Scientific Chemicals
H61302MD |
250 mg |
Each for $115.56
|
|
|||||
AAH6130203
|
Thermo Scientific Chemicals
H6130203 |
1 g |
Each for $369.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1427-06-1 | |
155.128 | |
HLYBWNNPVXFCPZ-UHFFFAOYSA-N | |
methyl 6-fluoropyridine-3-carboxylate |
C7H6FNO2 | |
MFCD03095098 | |
13758765 | |
COC(=O)C1=CN=C(C=C1)F |
Specifications
1427-06-1 | |
99°C (210°F) | |
MFCD03095098 | |
methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate | |
COC(=O)C1=CN=C(C=C1)F | |
155.128 | |
155.13 | |
Solid |
48°C to 52°C | |
C7H6FNO2 | |
250 mg | |
HLYBWNNPVXFCPZ-UHFFFAOYSA-N | |
methyl 6-fluoropyridine-3-carboxylate | |
13758765 | |
95% | |
Methyl 6-fluoronicotinate |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only