missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 6-Bromopyridine-2-carboxylate, TCI America™

$22.92 - $77.92

Chemical Identifiers

CAS 26218-75-7
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD06203934
InChI Key SGNCOKUHMXLGAH-UHFFFAOYSA-N
Synonym methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate
PubChem CID 16217886
IUPAC Name methyl 6-bromopyridine-2-carboxylate
SMILES COC(=O)C1=NC(=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23421G View Documents TCI America
M23421G
1g Each for $22.92
 
M23425G View Documents TCI America
M23425G
5g Each for $77.92
 
Specifications

Chemical Identifiers

26218-75-7
216.034
SGNCOKUHMXLGAH-UHFFFAOYSA-N
16217886
COC(=O)C1=NC(=CC=C1)Br
C7H6BrNO2
MFCD06203934
methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate
methyl 6-bromopyridine-2-carboxylate

Specifications

Undesignated
1g
C7H6BrNO2
methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate
COC(=O)C1=NC(=CC=C1)Br
216.034
216.03
≥98.0% (GC)
95°C
26218-75-7
MFCD06203934
SGNCOKUHMXLGAH-UHFFFAOYSA-N
methyl 6-bromopyridine-2-carboxylate
16217886
Crystal-Powder at 20°C
Methyl 6-Bromopyridine-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No