missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 6-Bromonicotinate, TCI America™

$22.92 - $68.75

Chemical Identifiers

CAS 26218-78-0
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD04972371
InChI Key NFLROFLPSNZIAH-UHFFFAOYSA-N
Synonym 6-Bromonicotinic Acid Methyl Ester, Methyl 6-Bromopyridine-3-carboxylate, 6-Bromopyridine-3-carboxylic Acid Methyl Ester
PubChem CID 13349623
IUPAC Name methyl 6-bromopyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26825G View Documents TCI America
M26825G
5g Each for $68.75
 
M26821G View Documents TCI America
M26821G
1g Each for $22.92
 
Specifications

Chemical Identifiers

26218-78-0
216.034
NFLROFLPSNZIAH-UHFFFAOYSA-N
13349623
COC(=O)C1=CN=C(C=C1)Br
C7H6BrNO2
MFCD04972371
6-Bromonicotinic Acid Methyl Ester, Methyl 6-Bromopyridine-3-carboxylate, 6-Bromopyridine-3-carboxylic Acid Methyl Ester
methyl 6-bromopyridine-3-carboxylate

Specifications

White
5g
C7H6BrNO2
6-Bromonicotinic Acid Methyl Ester, Methyl 6-Bromopyridine-3-carboxylate, 6-Bromopyridine-3-carboxylic Acid Methyl Ester
COC(=O)C1=CN=C(C=C1)Br
216.034
216.03
≥98.0% (GC)
108°C
26218-78-0
MFCD04972371
NFLROFLPSNZIAH-UHFFFAOYSA-N
methyl 6-bromopyridine-3-carboxylate
13349623
Crystal-Powder at 20°C
Methyl 6-Bromonicotinate
Safety and Handling

Safety and Handling

TSCA : No