missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-bromoindole-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H6318703

 View more versions of this product

Catalog No. AAH6318703



Description

Description

Applications
Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in acetone.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

Methyl 6-bromoindole-2-carboxylate
1g
C11H10BrNO2
ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester
CCOC(=O)C1=CC2=C(N1)C=C(C=C2)Br
268.11
254.08
97%
White
103858-53-3
MFCD03094942
FVMZWWFKRMBNSZ-UHFFFAOYSA-N
ethyl 6-bromo-1H-indole-2-carboxylate
7009496
Crystalline Powder

Chemical Identifiers

103858-53-3
268.11
FVMZWWFKRMBNSZ-UHFFFAOYSA-N
7009496
CCOC(=O)C1=CC2=C(N1)C=C(C=C2)Br
C11H10BrNO2
MFCD03094942
ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester
ethyl 6-bromo-1H-indole-2-carboxylate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents