Please login to your online account to display your discounted pricing

Methyl 6-bromohexanoate, +96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 6-bromohexanoate
Assay ≥96.0%
Assay Percent Range 96+%
Molecular Formula C7H13BrO2
Linear Formula Br(CH2)5C(O)OCH3
Formula Weight 209.08
MDL Number MFCD00187528
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392690050

  SDS
thermo fisher scientific
AC392690050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392690250

  SDS
thermo fisher scientific
AC392690250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392691000

  SDS
thermo fisher scientific
AC392691000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 209.08
Quantity 5g
Color Clear colorless to pale yellow
Density 1.316g/mL
Assay Percent Range 96+%
MDL Number MFCD00187528
CAS 14273-90-6
Chemical Name or Material Methyl 6-bromohexanoate
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4630 to 1.4650 (20°C, 589nm)
Molecular Formula C7H13BrO2
Linear Formula Br(CH2)5C(O)OCH3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.