missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-bromo-1-methylindole-2-carboxylate, 97%

$40.41 - $527.41

Chemical Identifiers

CAS 680569-18-0
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.11
MDL Number MFCD07364734
InChI Key RCDWEKOOOHATJC-UHFFFAOYSA-N
Synonym methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
PubChem CID 4777911
IUPAC Name methyl 6-bromo-1-methylindole-2-carboxylate
SMILES CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63655MD View Documents Alfa Aesar™
H63655MD
250mg Each for $40.41
 
AAH6365503 View Documents Alfa Aesar™
H6365503
1g Each for $127.23
 
AAH6365506 View Documents Alfa Aesar™
H6365506
5g Each for $527.41
 
Description

Description

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

680569-18-0
268.11
RCDWEKOOOHATJC-UHFFFAOYSA-N
4777911
CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC
C11H10BrNO2
MFCD07364734
methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
methyl 6-bromo-1-methylindole-2-carboxylate

Specifications

Yellow
680569-18-0
MFCD07364734
RCDWEKOOOHATJC-UHFFFAOYSA-N
methyl 6-bromo-1-methylindole-2-carboxylate
4777911
Solid
Methyl 6-bromo-1-methylindole-2-carboxylate
250mg
C11H10BrNO2
methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC
268.11
268.11
97%
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C