missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 6-bromo-1-methylindole-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H63655MD

 View more versions of this product

Catalog No. AAH63655MD



Description

Description

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Methyl 6-bromo-1-methylindole-2-carboxylate
250mg
C11H10BrNO2
methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC
268.11
268.11
97%
Yellow
680569-18-0
MFCD07364734
RCDWEKOOOHATJC-UHFFFAOYSA-N
methyl 6-bromo-1-methylindole-2-carboxylate
4777911
Solid

Chemical Identifiers

680569-18-0
268.11
RCDWEKOOOHATJC-UHFFFAOYSA-N
4777911
CN1C(=CC2=C1C=C(C=C2)Br)C(=O)OC
C11H10BrNO2
MFCD07364734
methyl 6-bromo-1-methyl-1h-indole-2-carboxylate, 6-bromo-1-methyl-1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, 6-bromo-1-methyl-, methyl ester
methyl 6-bromo-1-methylindole-2-carboxylate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents