missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-oxopyrrolidine-2-carboxylate, 95%

$74.23 - $878.91

Chemical Identifiers

CAS 54571-66-3
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.142
MDL Number MFCD03844651
InChI Key HQGPKMSGXAUKHT-UHFFFAOYSA-N
Synonym 5-oxo-2-pyrrolidinecarboxylic acid methyl ester, 5-oxo-dl-proline methyl ester, 5-oxo-pyrrolidine-2-carboxylic acid methyl ester, dl-pyroglutamic acid methyl ester, methyl 2-pyrolidone-5-carboxylate, methyl 5-oxo-2-pyrrolidinecarboxylate, methyl 5-oxo-2-pyrrolidinecarboxylate #, methyl 5-oxo-dl-prolinate, methyl dl-pyroglutamate, proline, 5-oxo-, methylester
PubChem CID 500249
IUPAC Name methyl 5-oxopyrrolidine-2-carboxylate
SMILES COC(=O)C1CCC(=O)N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64804MD View Documents Alfa Aesar™
H64804MD
250mg Each for $74.23
 
AAH6480403 View Documents Alfa Aesar™
H6480403
1g Each for $219.73
 
AAH6480406 View Documents Alfa Aesar™
H6480406
5g Each for $878.91
 
Specifications

Chemical Identifiers

54571-66-3
143.142
HQGPKMSGXAUKHT-UHFFFAOYSA-N
500249
COC(=O)C1CCC(=O)N1
C6H9NO3
MFCD03844651
5-oxo-2-pyrrolidinecarboxylic acid methyl ester, 5-oxo-dl-proline methyl ester, 5-oxo-pyrrolidine-2-carboxylic acid methyl ester, dl-pyroglutamic acid methyl ester, methyl 2-pyrolidone-5-carboxylate, methyl 5-oxo-2-pyrrolidinecarboxylate, methyl 5-oxo-2-pyrrolidinecarboxylate #, methyl 5-oxo-dl-prolinate, methyl dl-pyroglutamate, proline, 5-oxo-, methylester
methyl 5-oxopyrrolidine-2-carboxylate

Specifications

1.485
21°C to 23°C (literature)
54571-66-3
MFCD03844651
5-oxo-2-pyrrolidinecarboxylic acid methyl ester, 5-oxo-dl-proline methyl ester, 5-oxo-pyrrolidine-2-carboxylic acid methyl ester, dl-pyroglutamic acid methyl ester, methyl 2-pyrolidone-5-carboxylate, methyl 5-oxo-2-pyrrolidinecarboxylate, methyl 5-oxo-2-pyrrolidinecarboxylate #, methyl 5-oxo-dl-prolinate, methyl dl-pyroglutamate, proline, 5-oxo-, methylester
COC(=O)C1CCC(=O)N1
143.142
143.14
95%
133°C to 135°C (1mmHg)
250mg
C6H9NO3
81950
HQGPKMSGXAUKHT-UHFFFAOYSA-N
methyl 5-oxopyrrolidine-2-carboxylate
500249
Solid
Methyl 5-oxopyrrolidine-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 259-233-3

TSCA : No

Recommended Storage : Ambient temperatures