missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl-5-norbornene-2,3-dicarboxylic Anhydride (mixture of isomers) 80.0+%, TCI America™
Supplier: TCI America M0690500G
Specifications
Methyl-5-norbornene-2,3-dicarboxylic Anhydride (mixture of isomers) | |
Yellow | |
MFCD00167036 | |
3265 | |
KNRCVAANTQNTPT-UHFFFAOYNA-N | |
178.19 | |
Liquid |
25134-21-8 | |
C10H10O3 | |
500 g | |
Methyl-3,6-endomethylenetetrahydrophthalic Anhydride, Methylnadic Anhydride | |
178.19 | |
≥80.0% (GC) |
Chemical Identifiers
25134-21-8 | |
178.19 | |
KNRCVAANTQNTPT-UHFFFAOYNA-N |
C10H10O3 | |
MFCD00167036 | |
Methyl-3,6-endomethylenetetrahydrophthalic Anhydride, Methylnadic Anhydride |
Safety and Handling
EINECSNumber : (4)-0298
RTECSNumber : RB9100000
TSCA : Yes