missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers, ACROS Organics™
$75.75 - $75.75
Chemical Identifiers
CAS | 25134-21-8 |
---|---|
Molecular Formula | C10H10O3 |
Molecular Weight (g/mol) | 178.18 |
MDL Number | MFCD00167036 |
InChI Key | KNRCVAANTQNTPT-UHFFFAOYNA-N |
Synonym | Methyl nadic anhydride, Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC216270050
|
Thermo Fisher Scientific
216270050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC216272500
|
Thermo Fisher Scientific
216272500 |
250g | Glass bottle |
Each for $75.75
|
|
AC216270010
|
Thermo Fisher Scientific
216270010 |
1kg | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC216270051
|
Thermo Fisher Scientific
AC216270051 |
5kg | Plastic drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
25134-21-8 | |
178.18 | |
KNRCVAANTQNTPT-UHFFFAOYNA-N |
C10H10O3 | |
MFCD00167036 | |
Methyl nadic anhydride, Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride |
Specifications
1.2300g/mL | |
Authentic | |
1.5060 to 1.508 | |
220-300 mPa.s (25°C) | |
Yellow | |
25134-21-8 | |
C10H10O3 | |
17, 461 | |
Solubility in water: <1g/L | |
178.18 | |
Oily Liquid | |
Methyl-5-norbornene-2, 3-dicarboxylic anhydride, Mixture of isomers |
140°C | |
Glass bottle | |
1.23 | |
275°C | |
5g | |
94% min. (GC) | |
MFCD00167036 | |
Methyl nadic anhydride, Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride | |
KNRCVAANTQNTPT-UHFFFAOYNA-N | |
178.18 | |
95% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Toxic if inhaled.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes skin irritation
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 246-644-8