Please login to your online account to display your discounted pricing

Methyl 5-nitro-2-furoate, 97%, ACROS Organics™

$308.07 - $308.07

Chemical Identifiers

Chemical Name or Material Methyl 5-nitro-2-furoate
Name Note 97%
CAS 1874-23-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H5NO5
Formula Weight 171.11
MDL Number MFCD00051956
Synonym Methyl 5-nitrofuran-2-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC344520050

  SDS
thermo fisher scientific
344520050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC344520250

  SDS
thermo fisher scientific
344520250
Glass bottle 25g Each for $308.07
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 171.11
Quantity 5g
Color Yellow
Melting Point 78° to 83°C
RTECS Number LV2013000
Packaging Glass bottle
Synonym Methyl 5-nitrofuran-2-carboxylate
Chemical Name or Material Methyl 5-nitro-2-furoate
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6H5NO5
MDL Number MFCD00051956
CAS 1874-23-3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.