missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-methylsalicylate 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 133990050

 View more versions of this product

Catalog No. AC133990050


 

Specifications Description & Specifications

Specifications

≥96.0%
166.18
Clear slightly yellow
1.5320 to 1.5340 (20°C, 589nm)
22717-57-3
97%
CH3C6H3(OH)COOCH3
97%
Liquid
5g
Glass bottle
Authentic
MFCD00016485
C9H10O3
Methyl 5-methylsalicylate

Chemical Identifiers

22717-57-3
C9H10O3
166.18
Methyl 5-methylsalicylate, 97%
97%
CH3C6H3(OH)COOCH3
MFCD00016485
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 245-173-5

TSCA : TSCA