Please login to your online account to display your discounted pricing

Methyl 5-methylsalicylate 97%, ACROS Organics™

$59.91 - $59.91

Chemical Identifiers

Chemical Name or Material Methyl 5-methylsalicylate
Name Note 97%
CAS 22717-57-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O3
Linear Formula CH3C6H3(OH)COOCH3
Formula Weight 166.18
MDL Number MFCD00016485
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC133990050

  SDS
thermo fisher scientific
133990050
Glass bottle 5g Each for $59.91
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 166.18
Quantity 5g
Color Clear slightly yellow
EINECS Number 245-173-5
TSCA TSCA
Chemical Name or Material Methyl 5-methylsalicylate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C9H10O3
Linear Formula CH3C6H3(OH)COOCH3
MDL Number MFCD00016485
CAS 22717-57-3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.