missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-Methyl-2-nitrobenzoate 98.0+%, TCI America™

$169.59 - $580.56

Chemical Identifiers

CAS 20587-30-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00157275
InChI Key KFOICDVZQKFCGM-UHFFFAOYSA-N
Synonym 5-Methyl-2-nitrobenzoic Acid Methyl Ester, Methyl 6-Nitro-m-toluate, 6-Nitro-m-toluic Acid Methyl Ester
PubChem CID 3314122
IUPAC Name methyl 5-methyl-2-nitrobenzoate
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25445G View Documents TCI America
M25445G
5g Each for $169.59
 
M254425G View Documents TCI America
M254425G
25g Each for $580.56
 
Specifications

Chemical Identifiers

20587-30-8
195.174
KFOICDVZQKFCGM-UHFFFAOYSA-N
3314122
CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)OC
C9H9NO4
MFCD00157275
5-Methyl-2-nitrobenzoic Acid Methyl Ester, Methyl 6-Nitro-m-toluate, 6-Nitro-m-toluic Acid Methyl Ester
methyl 5-methyl-2-nitrobenzoate

Specifications

White-Yellow
5g
C9H9NO4
5-Methyl-2-nitrobenzoic Acid Methyl Ester, Methyl 6-Nitro-m-toluate, 6-Nitro-m-toluic Acid Methyl Ester
CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)OC
195.174
195.17
≥98.0% (GC)
80°C
20587-30-8
MFCD00157275
KFOICDVZQKFCGM-UHFFFAOYSA-N
methyl 5-methyl-2-nitrobenzoate
3314122
Crystal-Powder at 20°C
Methyl 5-Methyl-2-nitrobenzoate
Safety and Handling

Safety and Handling

TSCA : No