Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-methyl-2-furoate, 97%

$71.88 - $71.88

Chemical Identifiers

Chemical Name or Material Methyl 5-methyl-2-furoate
CAS 2527-96-0
Molecular Formula C7H8O3
Formula Weight 140.14
MDL Number MFCD04116374
Synonym 5-Methylfuran-2-carboxylic acid methyl ester; Methyl 5-methylfuran-2-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2757003

  SDS
alfa aesar™
H2757003
1g Each for $71.88
Description & Specifications

Specifications

Formula Weight 140.14
Quantity 1g
Flash Point 87°C (189°F)
Chemical Name or Material Methyl 5-methyl-2-furoate
Molecular Formula C7H8O3
CAS 2527-96-0
Health Hazard 1 Warning
MDL Number MFCD04116374
Assay 97%
Boiling Point 255°C
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Synonym 5-Methylfuran-2-carboxylic acid methyl ester; Methyl 5-methylfuran-2-carboxylate

Applications
Furans with suitable substituent in the 2- positions may be transformed into other systems with aromatic character with methoxylation of cyclic acetals of 1,4- dicarbonyl compounds, followed by intramolecular condensations in the acid solutions.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.