missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-Methoxysalicylate, TCI America™

$70.28 - $201.67

Chemical Identifiers

CAS 2905-82-0
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00017185
InChI Key DFNBGZODMHWKKK-UHFFFAOYSA-N
Synonym 5-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-5-methoxybenzoate, 2-Hydroxy-5-methoxybenzoic Acid Methyl Ester
PubChem CID 4072341
IUPAC Name methyl 2-hydroxy-5-methoxybenzoate
SMILES COC1=CC(=C(C=C1)O)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M212725G View Documents TCI America
M212725G*
25g Each for $201.67
 
M21275G View Documents TCI America
M21275G*
5g Each for $70.28
 
Specifications

Chemical Identifiers

2905-82-0
182.175
DFNBGZODMHWKKK-UHFFFAOYSA-N
4072341
COC1=CC(=C(C=C1)O)C(=O)OC
C9H10O4
MFCD00017185
5-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-5-methoxybenzoate, 2-Hydroxy-5-methoxybenzoic Acid Methyl Ester
methyl 2-hydroxy-5-methoxybenzoate

Specifications

240°C
25g
C9H10O4
5-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-5-methoxybenzoate, 2-Hydroxy-5-methoxybenzoic Acid Methyl Ester
COC1=CC(=C(C=C1)O)C(=O)OC
182.175
182.18
≥98.0% (GC)
Yellow
2905-82-0
MFCD00017185
DFNBGZODMHWKKK-UHFFFAOYSA-N
methyl 2-hydroxy-5-methoxybenzoate
4072341
Clear Liquid at 20°C
Methyl 5-Methoxysalicylate
Safety and Handling

Safety and Handling

TSCA : No