missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-Methoxy-3-oxovalerate, TCI America™

Manufacturer:  TCI America M08121G

 View more versions of this product

Catalog No. M08121G



Specifications

Specifications

Methyl 5-Methoxy-3-oxovalerate
Yellow
62462-05-9
MFCD00042840
OPURNNHYTATPKM-UHFFFAOYSA-N
methyl 5-methoxy-3-oxopentanoate
542416
Clear Liquid at 20°C
69°C
1g
C7H12O4
3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate
COCCC(=O)CC(=O)OC
160.169
160.17
≥96.0% (GC)

Chemical Identifiers

62462-05-9
160.169
OPURNNHYTATPKM-UHFFFAOYSA-N
542416
COCCC(=O)CC(=O)OC
C7H12O4
MFCD00042840
3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate
methyl 5-methoxy-3-oxopentanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents