Please login to your online account to display your discounted pricing

Methyl 5-iodosalicylate 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 5-iodosalicylate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7IO3
Linear Formula IC6H3(OH)COOCH3
Formula Weight 278.04
MDL Number MFCD00016462
Synonym Methyl 2-hydroxy-5-iodobenzoate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381770100

  SDS
fisher scientific acros
AC381770100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381770500

  SDS
fisher scientific acros
AC381770500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 278.04
Quantity 10g
Color White
Melting Point 73° to 78°C
Synonym Methyl 2-hydroxy-5-iodobenzoate
Linear Formula IC6H3(OH)COOCH3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material Methyl 5-iodosalicylate
CAS 4068-75-1
MDL Number MFCD00016462
Assay Percent Range 99%
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C8H7IO3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.