Please login to your online account to display your discounted pricing

α-Methyl-5-hydroxytryptamine maleate, Tocris

5-HT2B agonist

$123.83 - $520.12

Chemical Identifiers

Chemical Name or Material α-Methyl-5-hydroxytryptamine maleate
Assay Percent Range >99%
Molecular Formula C11H14N2O·C4H4O4
Formula Weight 306.32
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

055710

 
r & d systems
055710
10mg Each for $123.83

055750

 
r & d systems
055750
50mg Each for $520.12
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 306.32
Chemical Name or Material α-Methyl-5-hydroxytryptamine maleate
Assay Percent Range >99%
Molecular Formula C11H14N2O·C4H4O4
CAS 97469-12-0
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C