Please login to your online account to display your discounted pricing

α-Methyl-5-hydroxytryptamine maleate, Tocris


5-HT2B agonist

Quantity 10mg

Manufacturer: r & d systems  055710

 View more versions of this product

Catalog No. 055710

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 306.32
Molecular Formula C11H14N2O·C4H4O4
CAS 97469-12-0
Assay Percent Range >99%
Chemical Name or Material α-Methyl-5-hydroxytryptamine maleate
Recommended Storage Store at -20°C

Specifications

Chemical Name or Material α-Methyl-5-hydroxytryptamine maleate
Assay Percent Range >99%
Molecular Formula C11H14N2O·C4H4O4
Formula Weight 306.32
Accessories

Store at -20°C