missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-Hydroxynicotinate, TCI America™

$56.53 - $493.48

Chemical Identifiers

CAS 30766-22-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00191521
InChI Key KJJSHOHQQHACLE-UHFFFAOYSA-N
Synonym methyl 5-hydroxynicotinate, 5-hydroxynicotinic acid methyl ester, methyl5-hydroxynicotinate, methyl 5-hydroxy-3-pyridinecarboxylate, 5-hydroxy-nicotinic acid methyl ester, methyl-5-hydroxynicotinate, 5-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 5-hydroxy-, methyl ester, 5-hydroxy nicotinic acid methyl ester, acmc-1crme
PubChem CID 354317
IUPAC Name methyl 5-hydroxypyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H084125G View Documents TCI America
H084125G
25g Each for $493.48
 
H08415G View Documents TCI America
H08415G
5g Each for $165.00
 
H08411G View Documents TCI America
H08411G
1g Each for $56.53
 
Specifications

Chemical Identifiers

30766-22-4
153.137
KJJSHOHQQHACLE-UHFFFAOYSA-N
354317
COC(=O)C1=CC(=CN=C1)O
C7H7NO3
MFCD00191521
methyl 5-hydroxynicotinate, 5-hydroxynicotinic acid methyl ester, methyl5-hydroxynicotinate, methyl 5-hydroxy-3-pyridinecarboxylate, 5-hydroxy-nicotinic acid methyl ester, methyl-5-hydroxynicotinate, 5-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 5-hydroxy-, methyl ester, 5-hydroxy nicotinic acid methyl ester, acmc-1crme
methyl 5-hydroxypyridine-3-carboxylate

Specifications

White
25g
C7H7NO3
methyl 5-hydroxynicotinate, 5-hydroxynicotinic acid methyl ester, methyl5-hydroxynicotinate, methyl 5-hydroxy-3-pyridinecarboxylate, 5-hydroxy-nicotinic acid methyl ester, methyl-5-hydroxynicotinate, 5-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 5-hydroxy-, methyl ester, 5-hydroxy nicotinic acid methyl ester, acmc-1crme
COC(=O)C1=CC(=CN=C1)O
153.137
153.14
≥98.0% (GC,T)
190°C
30766-22-4
MFCD00191521
KJJSHOHQQHACLE-UHFFFAOYSA-N
methyl 5-hydroxypyridine-3-carboxylate
354317
Crystal-Powder at 20°C
Methyl 5-Hydroxynicotinate
Safety and Handling

Safety and Handling

TSCA : No