Please login to your online account to display your discounted pricing

Methyl 5-hydroxynicotinate, +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 5-hydroxynicotinate
CAS 30766-22-4
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C7H7NO3
Formula Weight 153.14
MDL Number MFCD00191521
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393352500

  SDS
thermo fisher scientific
AC393352500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393350050

  SDS
thermo fisher scientific
AC393350050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393350010

  SDS
thermo fisher scientific
AC393350010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.0%
Physical Form Powder and Lumps
Formula Weight 153.14
Quantity 250mg
Color Light beige
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 30766-22-4
Molecular Formula C7H7NO3
Assay Percent Range 97+%
Chemical Name or Material Methyl 5-hydroxynicotinate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00191521
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.