missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 5-Hydroxyanthranilate 98.0+%, TCI America™
$132.71 - $655.30
Chemical Identifiers
CAS | 1882-72-0 |
---|---|
Molecular Formula | C8H9NO3 |
Molecular Weight (g/mol) | 167.16 |
MDL Number | MFCD05863263 |
InChI Key | DWBKSTKVIIRFHL-UHFFFAOYSA-N |
Synonym | 5-Hydroxyanthranilic Acid Methyl Ester, Methyl 2-Amino-5-hydroxybenzoate, 2-Amino-5-hydroxybenzoic Acid Methyl Ester |
PubChem CID | 819940 |
IUPAC Name | methyl 2-amino-5-hydroxybenzoate |
SMILES | COC(=O)C1=C(N)C=CC(O)=C1 |
Chemical Identifiers
1882-72-0 | |
167.16 | |
DWBKSTKVIIRFHL-UHFFFAOYSA-N | |
819940 | |
COC(=O)C1=C(N)C=CC(O)=C1 |
C8H9NO3 | |
MFCD05863263 | |
5-Hydroxyanthranilic Acid Methyl Ester, Methyl 2-Amino-5-hydroxybenzoate, 2-Amino-5-hydroxybenzoic Acid Methyl Ester | |
methyl 2-amino-5-hydroxybenzoate |
Specifications
1882-72-0 | |
C8H9NO3 | |
1 g | |
DWBKSTKVIIRFHL-UHFFFAOYSA-N | |
methyl 2-amino-5-hydroxybenzoate | |
819940 | |
≥98.0% (GC,T) | |
Methyl 5-Hydroxyanthranilate |
160°C | |
MFCD05863263 | |
5-Hydroxyanthranilic Acid Methyl Ester, Methyl 2-Amino-5-hydroxybenzoate, 2-Amino-5-hydroxybenzoic Acid Methyl Ester | |
COC(=O)C1=C(N)C=CC(O)=C1 | |
167.16 | |
167.16 | |
Crystalline Powder |
Safety and Handling
TSCA : No