Learn More
Methyl 5-hexynoate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5337006
Description
Methyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.
Notes
Incompatible with oxidizing agents.
Specifications
Methyl 5-hexynoate | |
60°C to 62°C (10 mmHg) | |
HC≡C(CH2)3CO2CH3 | |
MFCD00671366 | |
methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester | |
COC(=O)CCCC#C | |
126.155 | |
126.16 |
77758-51-1 | |
C7H10O2 | |
1.434 | |
5 g | |
LZULAZTXJLWELL-UHFFFAOYSA-N | |
methyl hex-5-ynoate | |
534077 | |
97% |
Chemical Identifiers
77758-51-1 | |
126.155 | |
LZULAZTXJLWELL-UHFFFAOYSA-N | |
534077 | |
COC(=O)CCCC#C |
C7H10O2 | |
MFCD00671366 | |
methyl 5-hexynoate, 5-hexynoic acid, methyl ester, 5-hexynoic acid methyl ester, hex-5-ynoic acid methyl ester, methyl 5-hexynoate #, hex-5-ynoic acid, methyl ester | |
methyl hex-5-ynoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only