Please login to your online account to display your discounted pricing

Methyl 5-hexynoate 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC380880250

 View more versions of this product

Catalog No. AC380880250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 126.15
Quantity 25g
Color Clear colorless to pale yellow
Refractive Index 1.4330 to 1.4350 (20°C, 589nm)
Chemical Name or Material Methyl 5-hexynoate
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CHC(CH2)3COOCH3
CAS 77758-51-1
Assay Percent Range 95%
Molecular Formula C7H10O2
MDL Number MFCD00671366
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl 5-hexynoate
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H10O2
Linear Formula CHC(CH2)3COOCH3
Formula Weight 126.15
MDL Number MFCD00671366
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents