missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-Hexenoate, TCI America™

$259.73 - $776.13

Chemical Identifiers

CAS 2396-80-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00671538
InChI Key ASKDFGVMJZMYEM-UHFFFAOYSA-N
Synonym 5-Hexenoic Acid Methyl Ester
PubChem CID 520082
IUPAC Name methyl hex-5-enoate
SMILES COC(=O)CCCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H087225G View Documents TCI America
H087225G
25g Each for $776.13
 
H08725G View Documents TCI America
H08725G
5g Each for $259.73
 
Specifications

Chemical Identifiers

2396-80-7
128.171
ASKDFGVMJZMYEM-UHFFFAOYSA-N
520082
COC(=O)CCCC=C
C7H12O2
MFCD00671538
5-Hexenoic Acid Methyl Ester
methyl hex-5-enoate

Specifications

157°C
25g
C7H12O2
3272
ASKDFGVMJZMYEM-UHFFFAOYSA-N
methyl hex-5-enoate
520082
Clear Liquid at 20°C
Methyl 5-Hexenoate
Colorless
2396-80-7
MFCD00671538
5-Hexenoic Acid Methyl Ester
COC(=O)CCCC=C
128.171
128.17
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No