Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-fluoro-2-nitrobenzoate, 98%

Chemical Identifiers

Chemical Name or Material Methyl 5-fluoro-2-nitrobenzoate
CAS 393-85-1
Formula Weight 199.14
MDL Number MFCD03425619
Synonym 5-Fluoro-2-nitrobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6130706

  SDS
alfa aesar™
H6130706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6130714

  SDS
alfa aesar™
H6130714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 35° to 37°C
Formula Weight 199.14
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Chemical Name or Material Methyl 5-fluoro-2-nitrobenzoate
Synonym 5-Fluoro-2-nitrobenzoic acid methyl ester
Health Hazard 1 Warning
Physical Form Low Melting Solid
CAS 393-85-1
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD03425619

Applications
Methyl 5-fluoro-2-nitrobenzoate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.