Please login to your online account to display your discounted pricing

Methyl 5-(chlorosulfonyl)-2-furoate, 95+%, Maybridge

$88.65 - $88.65

Chemical Identifiers

Chemical Name or Material Methyl 5-(chlorosulfonyl)-2-furoate
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H5ClO5S
Formula Weight 224.62g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP01271CB

 
fisher scientific company
SP01271CB
Amber Glass Bottle 250mg Each for $88.65
Description & Specifications

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Chemical Name or Material Methyl 5-(chlorosulfonyl)-2-furoate
CAS 69816-05-3
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H5ClO5S
Formula Weight 224.62g/mol