Please login to your online account to display your discounted pricing

Methyl 5-(chlorosulfonyl)-2-furoate, 95+%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SP01271CB

 View more versions of this product

Catalog No. SP01271CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Packaging Amber Glass Bottle
Quantity 250mg
Chemical Name or Material Methyl 5-(chlorosulfonyl)-2-furoate
CAS 69816-05-3
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H5ClO5S
Formula Weight 224.62g/mol

Specifications

Chemical Name or Material Methyl 5-(chlorosulfonyl)-2-furoate
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H5ClO5S
Formula Weight 224.62g/mol