Please login to your online account to display your discounted pricing

Methyl 5-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 5-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate
CAS 306936-53-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H8ClNO4S
Formula Weight 237.66
MDL Number MFCD02180789
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP01464DA

  SDS
fisher scientific company
SP01464DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Formula Weight 237.66
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD02180789
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H8ClNO4S
Chemical Name or Material Methyl 5-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate
CAS 306936-53-8
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.