Please login to your online account to display your discounted pricing

Methyl 5-(chloromethyl)-2-furancarboxylate, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC367410050

 View more versions of this product

Catalog No. AC367410050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Low Melting Crystalline Solid or Oily Liquid
Formula Weight 174.58
Quantity 5g
Color White to yellow
Melting Point 31° to 35°C
Chemical Name or Material Methyl 5-(chloromethyl)-2-furancarboxylate
Assay Percent Range 99%
Molecular Formula C7H7ClO3
MDL Number MFCD00020922
CAS 2144-37-8
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C
Packaging Glass bottle

Specifications

Chemical Name or Material Methyl 5-(chloromethyl)-2-furancarboxylate
CAS 2144-37-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H7ClO3
Formula Weight 174.58
MDL Number MFCD00020922
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Refrigerator +4°C

SDS & Safety Documents