missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-(chloromethyl)-2-furancarboxylate, 99%, ACROS Organics™

Chemical Identifiers

CAS 2144-37-8
Molecular Formula C7H7ClO3
Molecular Weight (g/mol) 174.58
MDL Number MFCD00020922
InChI Key PWXMEBZOKUPCST-UHFFFAOYSA-N
Synonym 2-furancarboxylic acid, 5-chloromethyl-, methyl ester
PubChem CID 75065
IUPAC Name methyl 5-(chloromethyl)furan-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)CCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367410050 SDS View Documents Thermo Fisher Scientific
AC367410050
5g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

2144-37-8
174.58
PWXMEBZOKUPCST-UHFFFAOYSA-N
75065
COC(=O)C1=CC=C(O1)CCl
C7H7ClO3
MFCD00020922
2-furancarboxylic acid, 5-chloromethyl-, methyl ester
methyl 5-(chloromethyl)furan-2-carboxylate

Specifications

Authentic
White to Yellow
5g
99%
MFCD00020922
PWXMEBZOKUPCST-UHFFFAOYSA-N
methyl 5-(chloromethyl)furan-2-carboxylate
75065
Low Melting Crystalline Solid or Oily Liquid
Methyl 5-(chloromethyl)-2-furancarboxylate
Glass Bottle
31°C to 35°C
2144-37-8
C7H7ClO3
2-furancarboxylic acid, 5-chloromethyl-, methyl ester
COC(=O)C1=CC=C(O1)CCl
174.58
174.58
≥98.5%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Recommended Storage : Refrigerator +4°C