Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-chloro-2-nitrobenzoate, 98%

$360.12 - $360.12

Chemical Identifiers

Chemical Name or Material Methyl 5-chloro-2-nitrobenzoate
CAS 51282-49-6
Molecular Formula C8H6ClNO4
Formula Weight 215.59
MDL Number MFCD00272139
Synonym 5-Chloro-2-nitrobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1909814

  SDS
alfa aesar™
A1909814
25g Each for $360.12
Description & Specifications

Specifications

Formula Weight 215.59
Chemical Name or Material Methyl 5-chloro-2-nitrobenzoate
TSCA TSCA
Assay 98%
Molecular Formula C8H6ClNO4
MDL Number MFCD00272139
CAS 51282-49-6
Synonym 5-Chloro-2-nitrobenzoic acid methyl ester
Health Hazard 1 Warning
Melting Point 48° to 50°C
Quantity 25g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.