missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-chloro-2-hydroxybenzoate, 97%

$92.61 - $297.69

Chemical Identifiers

CAS 4068-78-4
Molecular Formula C8H7ClO3
Formula Weight 186.6
MDL Number MFCD00016461
Synonym 5-Chloro-2-hydroxybenzoic acid methyl ester; Methyl 5-chlorosalicylate
Chemical Name or Material Methyl 5-chloro-2-hydroxybenzoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1997188 SDS Alfa Aesar™
A1997188
2.5g Each for $92.61
AAA1997109 SDS Alfa Aesar™
A1997109
10g Each for $297.69
Specifications Description & Specifications

Chemical Identifiers

4068-78-4
186.6
5-Chloro-2-hydroxybenzoic acid methyl ester; Methyl 5-chlorosalicylate
C8H7ClO3
MFCD00016461
Methyl 5-chloro-2-hydroxybenzoate

Specifications

4068-78-4
186.6
5-Chloro-2-hydroxybenzoic acid methyl ester; Methyl 5-chlorosalicylate
2.5g
Methyl 5-chloro-2-hydroxybenzoate
C8H7ClO3
MFCD00016461
44° to 48°C
>110°C (230°F)
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA