Please login to your online account to display your discounted pricing

Methyl 5-bromovalerate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  126460250

 View more versions of this product

Catalog No. AC126460250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 195.06
Quantity 25g
Color Clear colorless to brown
TSCA TSCA
EINECS Number 226-709-7
Density 1.3600g/mL
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material Methyl 5-bromovalerate
Linear Formula Br(CH2)4CO2CH3
Molecular Formula C6H11BrO2
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00000265
CAS 5454-83-1

Specifications

Chemical Name or Material Methyl 5-bromovalerate
Name Note 97%
CAS 5454-83-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H11BrO2
Linear Formula Br(CH2)4CO2CH3
Formula Weight 195.06
MDL Number MFCD00000265